siRNA Details
Gene Symbol: ARL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103511.1 | 4 | 720 | |||
XM_006713783.3 | XP_006713846.1 | 3 | 2124 | |||
XM_017007311.1 | XP_016862800.1 | 5 | 2303 | |||
XM_017007312.1 | XP_016862801.1 | 4 | 2358 | |||
XM_017007313.1 | XP_016862802.1 | 4 | 2359 | |||
GenBank mRNA | AK292958.1 | - | 3 | 384 | ||
AK310829.1 | - | 3 | 375 | |||
AL136815.1 | - | 2 | 151 | |||
BC024239.2 | - | 4 | 490 | 4826873 | ||
BG722744.1 | - | 4 | 495 | |||
BM705017.1 | - | 4 | 341 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
