siRNA Details
Gene Symbol: GNLY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001302758.1 | NP_001289687.1 | 4 | 439 | ||
NM_006433.4 | NP_006424.2 | 3 | 358 | |||
NM_012483.3 | NP_036615.2 | 4 | 602 | |||
XM_005264084.2 | XP_005264141.1 | 4 | 372 | |||
XM_005264085.2 | XP_005264142.1 | 3 | 347 | |||
XM_005264087.2 | XP_005264144.1 | 4 | 599 | |||
GenBank mRNA | AK304728.1 | - | 4 | 372 | ||
AK308781.1 | - | 3 | 461 | |||
AK310057.1 | - | 4 | 600 | |||
AK311012.1 | - | 3 | 311 | |||
AK311537.1 | - | 4 | 492 | |||
AK311538.1 | - | 3 | 225 | |||
AK311657.1 | - | 3 | 888 | |||
BC023576.2 | - | 3 | 233 | 4766109 | ||
CR541859.1 | - | 3 | 230 | |||
X05044.1 | - | 4 | 462 | |||
X54101.1 | - | 3 | 358 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
