siRNA Details
Gene Symbol: GNLY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001302758.1 | NP_001289687.1 | 4, 5 | 462 | ||
NM_006433.4 | NP_006424.2 | 3, 4 | 381 | |||
NM_012483.3 | NP_036615.2 | 4, 5 | 625 | |||
GenBank mRNA | AK304728.1 | - | 4, 5 | 395 | ||
BC023576.2 | - | 3, 4 | 256 | 4766109 | ||
CR541859.1 | - | 3, 4 | 253 | |||
X05044.1 | - | 4, 5 | 485 | |||
X54101.1 | - | 3, 4 | 381 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
