siRNA Details
Gene Symbol: RGN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282848.1 | NP_001269777.1 | 8 | 2077 | ||
NM_001282849.1 | NP_001269778.1 | 7 | 1910 | |||
NM_004683.5 | NP_004674.1 | 7 | 1506 | |||
NM_152869.3 | NP_690608.1 | 8 | 2126 | |||
XM_006724567.2 | XP_006724630.1 | 6 | 1226 | |||
XM_006724568.2 | XP_006724631.1 | 5 | 1094 | |||
XM_017029954.1 | XP_016885443.1 | 6 | 1834 | |||
GenBank mRNA | AB028125.1 | - | 7 | 1230 | ||
AB032064.1 | - | 7 | 1288 | |||
AK223360.1 | - | 7 | 1506 | |||
AK290136.1 | - | 7 | 1444 | |||
BC050371.1 | - | 7 | 1848 | 5760604 | ||
BC073173.1 | - | 7 | 1320 | 6186669 | ||
D31815.1 | - | 7 | 1229 | |||
EU794655.1 | - | 7 | 1288 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
