siRNA Details
Gene Symbol: REPIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099695.1 | NP_001093165.1 | 3 | 3108 | ||
NM_001099696.2 | NP_001093166.1 | 2 | 2910 | |||
NM_013400.3 | NP_037532.2 | 4 | 3233 | |||
NM_014374.3 | NP_055189.2 | 1 | 2817 | |||
XM_005249985.1 | XP_005250042.1 | 4 | 3128 | |||
XM_006715947.3 | XP_006716010.1 | 3 | 3142 | |||
XM_006715948.3 | XP_006716011.1 | 3 | 3162 | |||
XM_006715949.3 | XP_006716012.1 | 3 | 3047 | |||
XM_006715950.3 | XP_006716013.1 | 3 | 3139 | |||
XM_006715951.2 | XP_006716014.1 | 3 | 3068 | |||
XM_006715952.2 | XP_006716015.1 | 4 | 3236 | |||
XM_006715953.2 | XP_006716016.1 | 2 | 2900 | |||
XM_011516112.1 | XP_011514414.1 | 2 | 3890 | |||
XM_017012081.1 | XP_016867570.1 | 3 | 3690 | |||
XM_017012082.1 | XP_016867571.1 | 4 | 3296 | |||
GenBank mRNA | AF201303.1 | - | 2 | 2868 | ||
AJ245553.1 | - | 1 | 2818 | |||
AK025356.1 | - | 1 | 2768 | |||
BC000363.2 | - | 1 | 2799 | 2964536 | ||
BC001760.1 | - | 1 | 2758 | 3353279 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
