siRNA Details
Gene Symbol: SNCA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000345.3 | NP_000336.1 | 6 | 674 | ||
NM_001146054.1 | NP_001139526.1 | 6 | 670 | |||
NM_001146055.1 | NP_001139527.1 | 6 | 481 | |||
NM_007308.2 | NP_009292.1 | 5 | 586 | |||
XM_011532208.2 | XP_011530510.1 | 6 | 551 | |||
XM_017008562.1 | XP_016864051.1 | 6 | 545 | |||
XM_017008563.1 | XP_016864052.1 | 6 | 571 | |||
GenBank mRNA | AK290169.1 | - | 6 | 495 | ||
AY049786.1 | - | 5 | 413 | |||
BC013293.2 | - | 6 | 834 | 3604532 | ||
BC108275.1 | - | 6 | 628 | 6147966 | ||
D31839.1 | - | 4 | 340 | |||
DA391115.1 | - | 6 | 481 | |||
JN709859.1 | - | 5 | 628 | |||
JN709860.1 | - | 4 | 544 | |||
JN709861.1 | - | 5 | 590 | |||
JN709862.1 | - | 5 | 632 | |||
JN709863.1 | - | 4 | 548 | |||
JN709864.1 | - | 5 | 397 | |||
JN709865.1 | - | 5 | 439 | |||
JN709866.1 | - | 4 | 355 | |||
JN709867.1 | - | 5 | 439 | |||
JN709868.1 | - | 5 | 481 | |||
JN709869.1 | - | 4 | 397 | |||
L08850.1 | - | 6 | 463 | |||
L36674.1 | - | 4 | 387 | |||
L36675.1 | - | 4 | 345 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
