siRNA Details
Gene Symbol: SNCA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000345.3 | NP_000336.1 | 4 | 546 | ||
NM_001146054.1 | NP_001139526.1 | 4 | 542 | |||
NM_001146055.1 | NP_001139527.1 | 4 | 353 | |||
NM_007308.2 | NP_009292.1 | 4 | 542 | |||
XM_011532203.1 | XP_011530505.1 | 4 | 575 | |||
XM_011532204.2 | XP_011530506.1 | 4 | 679 | |||
XM_011532205.2 | XP_011530507.1 | 4 | 381 | |||
XM_011532206.1 | XP_011530508.1 | 4 | 403 | |||
XM_011532207.1 | XP_011530509.1 | 4 | 390 | |||
XM_011532208.2 | XP_011530510.1 | 4 | 423 | |||
XM_017008562.1 | XP_016864051.1 | 4 | 417 | |||
XM_017008563.1 | XP_016864052.1 | 4 | 443 | |||
GenBank mRNA | AK290169.1 | - | 4 | 367 | ||
AY049786.1 | - | 3 | 285 | |||
BC013293.2 | - | 4 | 706 | 3604532 | ||
BC108275.1 | - | 4 | 500 | 6147966 | ||
CR457058.1 | - | 3 | 283 | |||
CR541653.1 | - | 3 | 283 | |||
D31839.1 | - | 3 | 296 | |||
DA391115.1 | - | 4 | 353 | |||
JN709859.1 | - | 3 | 500 | |||
JN709860.1 | - | 3 | 500 | |||
JN709861.1 | - | 4 | 546 | |||
JN709862.1 | - | 3 | 504 | |||
JN709863.1 | - | 3 | 504 | |||
JN709864.1 | - | 4 | 353 | |||
JN709865.1 | - | 3 | 311 | |||
JN709866.1 | - | 3 | 311 | |||
JN709867.1 | - | 4 | 395 | |||
JN709868.1 | - | 3 | 353 | |||
JN709869.1 | - | 3 | 353 | |||
L08850.1 | - | 4 | 335 | |||
L36674.1 | - | 2 | 259 | |||
L36675.1 | - | 3 | 301 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
