siRNA Details
Gene Symbol: NCF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000631.4 | NP_000622.2 | 2 | 257 | ||
NM_013416.3 | NP_038202.2 | 2 | 257 | |||
XM_017028808.1 | XP_016884297.1 | 2 | 621 | |||
GenBank mRNA | AB025219.1 | - | 2 | 109 | ||
AB025220.1 | - | 2 | 109 | |||
AK223324.1 | - | 2 | 228 | |||
AK290924.1 | - | 2 | 179 | |||
BC002798.1 | - | 2 | 257 | 3634177 | ||
BT007346.1 | - | 2 | 73 | |||
BX355636.2 | - | 2 | 233 | |||
CR456528.1 | - | 2 | 154 | |||
CR542078.1 | - | 2 | 73 | |||
X77094.1 | - | 2 | 203 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
