siRNA Details
Gene Symbol: UNC45A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039675.1 | NP_001034764.1 | 6 | 1030 | ||
NM_001323619.1 | NP_001310548.1 | 3 | 289 | |||
NM_001323620.1 | NP_001310549.1 | 3 | 679 | |||
NM_001323621.1 | NP_001310550.1 | 3 | 634 | |||
NM_018671.4 | NP_061141.2 | 3 | 679 | |||
XM_005254963.3 | XP_005255020.1 | 3 | 251 | |||
XM_011521779.2 | XP_011520081.1 | 6 | 1275 | |||
XM_011521780.2 | XP_011520082.1 | 2 | 181 | |||
XM_017022407.1 | XP_016877896.1 | 3 | 251 | |||
GenBank mRNA | AB014729.1 | - | 3 | 221 | ||
AB014736.1 | - | 3 | 263 | |||
AK000905.1 | - | 3 | 255 | |||
AK125721.1 | - | 6 | 1030 | |||
AK291622.1 | - | 3 | 234 | |||
AK293538.1 | - | 3 | 255 | |||
AK296964.1 | - | 3 | 256 | |||
AK302979.1 | - | 3 | 296 | |||
BC006214.1 | - | 3 | 262 | 3051384 | ||
BC037992.1 | - | 3 | 262 | 5534906 | ||
BC045635.1 | - | 6 | 1030 | 5297091 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
