siRNA Details
Gene Symbol: UNC45A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039675.1 | NP_001034764.1 | 5 | 930 | ||
NM_001323619.1 | NP_001310548.1 | 2 | 189 | |||
NM_001323620.1 | NP_001310549.1 | 2 | 579 | |||
NM_001323621.1 | NP_001310550.1 | 2 | 534 | |||
NM_018671.4 | NP_061141.2 | 2 | 579 | |||
XM_005254963.3 | XP_005255020.1 | 2 | 151 | |||
XM_011521779.2 | XP_011520081.1 | 5 | 1175 | |||
XM_011521780.2 | XP_011520082.1 | 1 | 81 | |||
XM_017022407.1 | XP_016877896.1 | 2 | 151 | |||
GenBank mRNA | AB014729.1 | - | 2 | 121 | ||
AB014736.1 | - | 2 | 163 | |||
AK000905.1 | - | 2 | 155 | |||
AK125721.1 | - | 5 | 930 | |||
AK291622.1 | - | 2 | 134 | |||
AK293538.1 | - | 2 | 155 | |||
AK296964.1 | - | 2 | 135 | |||
AK302979.1 | - | 2 | 196 | |||
BC006214.1 | - | 2 | 162 | 3051384 | ||
BC037992.1 | - | 2 | 162 | 5534906 | ||
BC045635.1 | - | 5 | 930 | 5297091 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
