siRNA Details
Gene Symbol: SCARA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016240.2 | NP_057324.2 | 5 | 1109 | ||
NM_182826.1 | NP_878185.1 | 5 | 1109 | |||
XM_017013535.1 | XP_016869024.1 | 6 | 1156 | |||
XM_017013536.1 | XP_016869025.1 | 5 | 1280 | |||
XM_017013537.1 | XP_016869026.1 | 5 | 1280 | |||
GenBank mRNA | AB007829.1 | - | 5 | 1109 | ||
AB007830.1 | - | 5 | 1109 | |||
BC060811.1 | - | 5 | 1301 | 30335021 | ||
BC075067.3 | - | 5 | 988 | 30915376 | ||
BC075068.2 | - | 5 | 988 | 30915589 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
