siRNA Details
Gene Symbol: TNFSF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003701.3 | NP_003692.1 | 5 | 1253 | ||
NM_033012.3 | NP_143026.1 | 7 | 1414 | |||
XM_011535280.2 | XP_011533582.1 | 5 | 958 | |||
XM_017020802.1 | XP_016876291.1 | 5 | 1060 | |||
XM_017020803.1 | XP_016876292.1 | 5 | 907 | |||
GenBank mRNA | AF013171.1 | - | 4 | 887 | ||
AF019047.1 | - | 5 | 1232 | |||
AF053712.1 | - | 5 | 1288 | |||
BC040889.1 | - | 7 | 1414 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
