siRNA Details
Gene Symbol: TNFSF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003701.3 | NP_003692.1 | 5 | 1057 | ||
NM_033012.3 | NP_143026.1 | 7 | 1218 | |||
XM_011535280.2 | XP_011533582.1 | 5 | 762 | |||
XM_017020802.1 | XP_016876291.1 | 5 | 864 | |||
XM_017020803.1 | XP_016876292.1 | 5 | 711 | |||
GenBank mRNA | AB037599.1 | - | 5 | 783 | ||
AB061227.1 | - | 6 | 865 | |||
AB064268.1 | - | 6 | 772 | |||
AB064269.1 | - | 5 | 988 | |||
AB064270.1 | - | 6 | 926 | |||
AF013171.1 | - | 4 | 692 | |||
AF019047.1 | - | 5 | 1036 | |||
AF053712.1 | - | 5 | 1092 | |||
BC040889.1 | - | 7 | 1218 | |||
BC074823.2 | - | 5 | 938 | 30915538 | ||
BC074890.2 | - | 5 | 938 | 30915245 | ||
BC117286.1 | - | 5 | 940 | 40125837 | ||
BC117288.1 | - | 5 | 940 | 40125839 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
