siRNA Details
Gene Symbol: ARMCX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016607.3 | NP_057691.1 | 5 | 2208 | ||
NM_177947.2 | NP_808816.1 | 5 | 2180 | |||
NM_177948.2 | NP_808817.1 | 5 | 2131 | |||
XM_005262141.2 | XP_005262198.1 | 5 | 2337 | |||
GenBank mRNA | BX647802.1 | - | 5 | 2149 | ||
CR933656.1 | - | 5 | 2302 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
