siRNA Details
Gene Symbol: KIAA1958 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287036.1 | NP_001273965.1 | 5 | 2685 | ||
NM_001287038.1 | NP_001273967.1 | 4 | 2591 | |||
NM_133465.3 | NP_597722.1 | 4 | 2601 | |||
GenBank mRNA | AB075838.1 | - | 3 | 2101 | ||
AK056181.1 | - | 3 | 1813 | |||
AK096613.1 | - | 4 | 2602 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
