siRNA Details
Gene Symbol: DVL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004421.2 | NP_004412.2 | 15 | 2456 | ||
XM_005244731.3 | XP_005244788.1 | 15 | 2561 | |||
GenBank mRNA | AB209210.1 | - | 9 | 3940 | ||
AF006011.1 | - | 15 | 2412 | |||
AK093189.1 | - | 8 | 1696 | |||
AK095867.1 | - | 8 | 1674 | |||
BC017225.2 | - | 13 | 2181 | |||
BC050454.1 | - | 9 | 1801 | 6201003 | ||
BC111419.1 | - | 1 | 275 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
