siRNA Details
Gene Symbol: HOPX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145459.1 | NP_001138931.1 | 2 | 288 | ||
NM_001145460.1 | NP_001138932.1 | 3 | 642 | |||
NM_032495.5 | NP_115884.4 | 3 | 642 | |||
NM_139211.4 | NP_631957.1 | 2 | 244 | |||
NM_139212.3 | NP_631958.1 | 2 | 517 | |||
XM_006714052.2 | XP_006714115.1 | 3 | 637 | |||
XM_017008728.1 | XP_016864217.1 | 2 | 279 | |||
XM_017008729.1 | XP_016864218.1 | 2 | 161 | |||
XM_017008730.1 | XP_016864219.1 | 2 | 209 | |||
XM_017008731.1 | XP_016864220.1 | 2 | 244 | |||
XM_017008732.1 | XP_016864221.1 | 3 | 520 | |||
XM_017008733.1 | XP_016864222.1 | 2 | 84 | |||
XM_017008734.1 | XP_016864223.1 | 2 | 140 | |||
GenBank mRNA | AB059408.1 | - | 2 | 202 | ||
AB059409.1 | - | 2 | 85 | |||
AB059410.2 | - | 2 | 94 | |||
AF454763.1 | - | 2 | 107 | |||
AF492675.1 | - | 2 | 491 | |||
AF492676.1 | - | 3 | 223 | |||
AF492677.1 | - | 3 | 267 | |||
AF492678.1 | - | 3 | 267 | |||
AF492679.1 | - | 3 | 223 | |||
AF492680.1 | - | 2 | 243 | |||
AF492681.1 | - | 2 | 206 | |||
AK289707.1 | - | 2 | 186 | |||
AW955211.1 | - | 2 | 210 | |||
BC014225.2 | - | 1 | 357 | 4335211 | ||
CB996541.1 | - | 2 | 280 | |||
CK002567.1 | - | 2 | 120 | |||
DA512059.1 | - | 2 | 517 | |||
EB386856.1 | - | 2 | 238 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
