siRNA Details
Gene Symbol: ENOX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281736.1 | NP_001268665.1 | 12 | 1847 | ||
NM_006375.3 | NP_006366.2 | 13 | 1991 | |||
NM_182314.2 | NP_872114.1 | 14 | 2080 | |||
XM_005262354.3 | XP_005262411.1 | 13 | 1936 | |||
XM_011531244.2 | XP_011529546.1 | 14 | 2082 | |||
XM_011531245.2 | XP_011529547.1 | 14 | 2076 | |||
XM_011531247.2 | XP_011529549.1 | 15 | 2208 | |||
XM_011531249.2 | XP_011529551.1 | 12 | 1636 | |||
XM_011531251.2 | XP_011529553.1 | 12 | 1741 | |||
XM_017029226.1 | XP_016884715.1 | 16 | 2299 | |||
XM_017029227.1 | XP_016884716.1 | 14 | 2033 | |||
XM_017029228.1 | XP_016884717.1 | 13 | 1782 | |||
XM_017029229.1 | XP_016884718.1 | 13 | 1985 | |||
GenBank mRNA | AF207881.2 | - | 11 | 1601 | ||
AK000353.1 | - | 12 | 1764 | |||
AK289812.1 | - | 14 | 1972 | |||
AK289837.1 | - | 13 | 1883 | |||
BC019254.1 | - | 8 | 1116 | |||
BC139781.1 | - | 11 | 1664 | |||
BC140874.1 | - | 12 | 1761 | 9021742 | ||
S72904.1 | - | 6 | 757 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
