siRNA Details
Gene Symbol: ENOX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281736.1 | NP_001268665.1 | 12 | 1872 | ||
NM_006375.3 | NP_006366.2 | 13 | 2016 | |||
NM_182314.2 | NP_872114.1 | 14 | 2105 | |||
XM_005262354.3 | XP_005262411.1 | 13 | 1961 | |||
XM_011531244.2 | XP_011529546.1 | 14 | 2107 | |||
XM_011531245.2 | XP_011529547.1 | 14 | 2101 | |||
XM_011531247.2 | XP_011529549.1 | 15 | 2233 | |||
XM_011531249.2 | XP_011529551.1 | 12 | 1661 | |||
XM_011531251.2 | XP_011529553.1 | 12 | 1766 | |||
XM_017029226.1 | XP_016884715.1 | 16 | 2324 | |||
XM_017029227.1 | XP_016884716.1 | 14 | 2058 | |||
XM_017029228.1 | XP_016884717.1 | 13 | 1807 | |||
XM_017029229.1 | XP_016884718.1 | 13 | 2010 | |||
GenBank mRNA | AF207881.2 | - | 11 | 1626 | ||
AK000353.1 | - | 12 | 1789 | |||
AK289812.1 | - | 14 | 1997 | |||
AK289837.1 | - | 13 | 1908 | |||
BC019254.1 | - | 8 | 1141 | |||
BC139781.1 | - | 11 | 1689 | |||
BC140874.1 | - | 12 | 1786 | 9021742 | ||
S72904.1 | - | 6 | 782 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
