siRNA Details
Gene Symbol: ENOX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281736.1 | NP_001268665.1 | 14 | 3542 | ||
NM_006375.3 | NP_006366.2 | 15 | 3686 | |||
NM_182314.2 | NP_872114.1 | 16 | 3775 | |||
XM_011531244.2 | XP_011529546.1 | 16 | 3777 | |||
XM_011531245.2 | XP_011529547.1 | 16 | 3771 | |||
XM_017029229.1 | XP_016884718.1 | 15 | 3680 | |||
GenBank mRNA | AF207881.2 | - | 13 | 3296 | ||
BC140874.1 | - | 14 | 3456 | 9021742 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
