siRNA Details
Gene Symbol: XAGE1E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001097604.2 | NP_001091073.2 | 2 | 317 | ||
NM_001097605.2 | NP_001091074.1 | 2 | 164 | |||
NR_033257.1 | 2 | 428 | ||||
XM_017029746.1 | XP_016885235.1 | 2 | 433 | |||
XM_017029747.1 | XP_016885236.1 | 2 | 317 | |||
GenBank mRNA | AF251237.1 | - | 2 | 309 | ||
AJ290447.1 | - | 2 | 153 | |||
AJ318878.1 | - | 2 | 428 | |||
AJ318879.1 | - | 2 | 153 | |||
BC009538.1 | - | 2 | 212 | |||
BT007099.1 | - | 1 | 29 |
Gene Symbol: XAGE1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001097594.2 | NP_001091063.2 | 2 | 317 | ||
NM_001097596.2 | NP_001091065.1 | 2 | 164 | |||
NR_033254.1 | 2 | 428 | ||||
XM_017029748.1 | XP_016885237.1 | 2 | 433 | |||
XM_017029749.1 | XP_016885238.1 | 2 | 317 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
