siRNA Details
Gene Symbol: LRFN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020737.2 | NP_065788.1 | 2 | 868 | ||
XM_011514761.2 | XP_011513063.1 | 3 | 696 | |||
XM_011514762.2 | XP_011513064.1 | 2 | 436 | |||
XM_017011110.1 | XP_016866599.1 | 3 | 1737 | |||
XM_017011111.1 | XP_016866600.1 | 3 | 694 | |||
GenBank mRNA | AB033072.1 | - | 2 | 791 | ||
AK291474.1 | - | 2 | 844 | |||
BC142616.1 | - | 2 | 777 | 40147341 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
