siRNA Details
Gene Symbol: CPM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005502.2 | NP_001005502.1 | 9 | 2113 | ||
NM_001874.4 | NP_001865.1 | 9 | 2145 | |||
NM_198320.3 | NP_938079.1 | 9 | 2131 | |||
GenBank mRNA | AF368463.1 | - | 9 | 2103 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
