siRNA Details
Gene Symbol: FOCAD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017794.4 | NP_060264.4 | 38 | 4615 | ||
XM_005251494.3 | XP_005251551.2 | 36 | 4599 | |||
XM_011517945.2 | XP_011516247.1 | 35 | 4203 | |||
XM_017014852.1 | XP_016870341.1 | 37 | 4389 | |||
XM_017014853.1 | XP_016870342.1 | 36 | 4284 | |||
XM_017014854.1 | XP_016870343.1 | 36 | 4284 | |||
XM_017014855.1 | XP_016870344.1 | 37 | 4488 | |||
XM_017014856.1 | XP_016870345.1 | 37 | 4429 | |||
XM_017014857.1 | XP_016870346.1 | 35 | 4494 | |||
XM_017014858.1 | XP_016870347.1 | 35 | 4494 | |||
XM_017014859.1 | XP_016870348.1 | 32 | 4035 | |||
GenBank mRNA | AB058700.1 | - | 24 | 2721 | ||
AK000382.1 | - | 4 | 339 | |||
AK055116.1 | - | 17 | 1923 | |||
AK172818.1 | - | 10 | 1174 | |||
AK225483.1 | - | 4 | 339 | |||
AY139834.1 | - | 38 | 4614 | |||
BC001246.1 | - | 4 | 355 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
