siRNA Details
Gene Symbol: FOCAD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017794.4 | NP_060264.4 | 43 | 5340 | ||
XM_005251494.3 | XP_005251551.2 | 41 | 5324 | |||
XM_011517945.2 | XP_011516247.1 | 40 | 4928 | |||
XM_017014852.1 | XP_016870341.1 | 42 | 5114 | |||
XM_017014853.1 | XP_016870342.1 | 41 | 5009 | |||
XM_017014854.1 | XP_016870343.1 | 41 | 5009 | |||
XM_017014855.1 | XP_016870344.1 | 42 | 5213 | |||
XM_017014856.1 | XP_016870345.1 | 42 | 5154 | |||
XM_017014857.1 | XP_016870346.1 | 40 | 5219 | |||
XM_017014858.1 | XP_016870347.1 | 40 | 5219 | |||
XM_017014859.1 | XP_016870348.1 | 37 | 4760 | |||
GenBank mRNA | AB058700.1 | - | 29 | 3446 | ||
AK000382.1 | - | 9 | 1062 | |||
AK055116.1 | - | 22 | 2648 | |||
AK172818.1 | - | 15 | 1899 | |||
AK225483.1 | - | 9 | 1062 | |||
AY139834.1 | - | 43 | 5339 | |||
BC001246.1 | - | 9 | 1080 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
