siRNA Details
Gene Symbol: OTOA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161683.1 | NP_001155155.1 | 22 | 2926 | ||
NM_144672.3 | NP_653273.3 | 26 | 3070 | |||
NM_170664.2 | NP_733764.1 | 17 | 2164 | |||
XM_011545747.2 | XP_011544049.1 | 27 | 3449 | |||
XM_011545748.2 | XP_011544050.2 | 16 | 2443 | |||
XM_017022950.1 | XP_016878439.1 | 27 | 3173 | |||
XM_017022951.1 | XP_016878440.1 | 12 | 1467 | |||
GenBank mRNA | AK093062.1 | - | 17 | 2164 | ||
AK125840.1 | - | 22 | 2926 | |||
AK302733.1 | - | 16 | 2226 | |||
BC038458.1 | - | 10 | 1296 | |||
BC129992.1 | - | 26 | 3067 |
Gene Symbol: LOC653786 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003676.3 | 7 | 2127 | |||
GenBank mRNA | AK092323.1 | - | 7 | 2127 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
