siRNA Details
Gene Symbol: OTOA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161683.1 | NP_001155155.1 | 20 | 2743 | ||
NM_144672.3 | NP_653273.3 | 24 | 2887 | |||
NM_170664.2 | NP_733764.1 | 15 | 1981 | |||
XM_011545747.2 | XP_011544049.1 | 25 | 3266 | |||
XM_011545748.2 | XP_011544050.2 | 14 | 2260 | |||
XM_017022950.1 | XP_016878439.1 | 25 | 2990 | |||
XM_017022951.1 | XP_016878440.1 | 10 | 1284 | |||
GenBank mRNA | AK093062.1 | - | 15 | 1981 | ||
AK125840.1 | - | 20 | 2743 | |||
AK302733.1 | - | 14 | 2043 | |||
BC038458.1 | - | 8 | 1113 | |||
BC129992.1 | - | 24 | 2884 | |||
BC129993.1 | - | 24 | 2891 | 40128985 |
Gene Symbol: LOC653786 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003676.3 | 5 | 1944 | |||
GenBank mRNA | AK092323.1 | - | 5 | 1944 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
