siRNA Details
Gene Symbol: MAFF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161572.1 | NP_001155044.1 | 3 | 1229 | ||
NM_001161573.1 | NP_001155045.1 | 3 | 987 | |||
NM_001161574.1 | NP_001155046.1 | 2 | 1136 | |||
NM_012323.3 | NP_036455.1 | 3 | 1203 | |||
GenBank mRNA | AB025247.1 | - | 3 | 1037 | ||
AJ010857.1 | - | 3 | 1124 | |||
AK002001.1 | - | 3 | 1144 | |||
AK097953.1 | - | 3 | 1094 | |||
BC015037.1 | - | 3 | 1076 | 3922552 | ||
BC067751.1 | - | 3 | 1142 | 30335753 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
