siRNA Details
Gene Symbol: ARID2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152641.2 | NP_689854.2 | 15 | 2943 | ||
XM_006719272.3 | XP_006719335.1 | 15 | 3097 | |||
XM_011538025.1 | XP_011536327.1 | 9 | 2050 | |||
GenBank mRNA | AK027718.1 | - | 1 | 97 | ||
AK127872.1 | - | 6 | 764 | |||
AL832200.1 | - | 11 | 2496 | |||
AY727870.2 | - | 15 | 2943 | |||
BC090062.1 | - | 1 | 18 | |||
BX537879.1 | - | 1 | 571 | |||
BX648152.1 | - | 9 | 2231 | |||
BX649133.1 | - | 7 | 1789 | |||
CR749833.1 | - | 14 | 2862 | |||
DQ096628.1 | - | 15 | 2943 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
