siRNA Details
Gene Symbol: ARID2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152641.2 | NP_689854.2 | 15 | 4191 | ||
XM_006719272.3 | XP_006719335.1 | 15 | 4345 | |||
XM_011538025.1 | XP_011536327.1 | 9 | 3298 | |||
GenBank mRNA | AB046777.1 | - | 1 | 1104 | ||
AK027718.1 | - | 1 | 1345 | |||
AK075385.1 | - | 1 | 745 | |||
AK127872.1 | - | 6 | 2012 | |||
AL832200.1 | - | 11 | 3744 | |||
BC090062.1 | - | 1 | 1266 | |||
BX537879.1 | - | 1 | 1819 | |||
BX648152.1 | - | 9 | 3479 | |||
BX649133.1 | - | 7 | 3037 | |||
CR749833.1 | - | 14 | 4110 | |||
DQ096628.1 | - | 15 | 4191 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
