siRNA Details
Gene Symbol: RSU1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012425.3 | NP_036557.1 | 9 | 940 | ||
NM_152724.2 | NP_689937.2 | 8 | 828 | |||
XM_005252552.3 | XP_005252609.1 | 8 | 890 | |||
GenBank mRNA | AK292911.1 | - | 8 | 753 | ||
AK312520.1 | - | 9 | 883 | |||
BC005993.1 | - | 9 | 910 | 4050837 | ||
BC008691.1 | - | 8 | 1099 | 3872071 | ||
BC015644.2 | - | 9 | 900 | 4647836 | ||
BC107898.1 | - | 8 | 807 | |||
BX647733.1 | - | 8 | 804 | |||
CR536521.1 | - | 8 | 797 | |||
CR541840.1 | - | 8 | 797 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
