siRNA Details
Gene Symbol: RSU1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012425.3 | NP_036557.1 | 9 | 2004 | ||
NM_152724.2 | NP_689937.2 | 8 | 1892 | |||
XM_005252552.3 | XP_005252609.1 | 8 | 1954 | |||
GenBank mRNA | AK055596.1 | - | 1 | 515 | ||
BX647733.1 | - | 8 | 1867 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
