siRNA Details
Gene Symbol: PCDH8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002590.3 | NP_002581.2 | 3 | 3556 | ||
NM_032949.2 | NP_116567.1 | 3 | 3265 | |||
GenBank mRNA | AF061573.2 | - | 3 | 3555 | ||
AK293747.1 | - | 4 | 3085 | |||
AK297652.1 | - | 4 | 2134 | |||
BC036025.1 | - | 3 | 3556 | 5278690 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
