siRNA Details
Gene Symbol: LOC401357 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130727.1 | 1 | 1218 | |||
GenBank mRNA | AK125790.1 | - | 1 | 1217 | ||
AK128114.1 | - | 1 | 959 | |||
BC137417.1 | - | 1 | 953 |
Gene Symbol: LOC100132217 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK128836.1 | - | 1 | 232 |
Gene Symbol: LOC107986803 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017012930.1 | XP_016868419.1 | 1 | 181 |
Gene Symbol: LOC107986804 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017012931.1 | XP_016868420.1 | 1 | 146 |
Gene Symbol: LOC107986805 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017012932.1 | XP_016868421.1 | 1 | 332 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
