siRNA Details
Gene Symbol: HLA-DRB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC001023.2 | - | 6 | 947 | 3345335 | |
M16941.1 | - | 6 | 909 |
Gene Symbol: HLA-DRB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC024269.1 | - | 6 | 930 | 4852401 | |
L42143.1 | - | 6 | 887 | |||
M14661.1 | - | 5 | 749 | |||
M14662.1 | - | 5 | 800 | |||
X02902.1 | - | 6 | 943 |
Gene Symbol: HLA-DRB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK292140.1 | - | 6 | 950 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
