siRNA Details
Gene Symbol: NEDD9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142393.1 | NP_001135865.1 | 3 | 512 | ||
NM_006403.3 | NP_006394.1 | 2 | 317 | |||
NM_182966.3 | NP_892011.2 | 2 | 317 | |||
NR_073131.1 | 4 | 606 | ||||
GenBank mRNA | AK292682.1 | - | 3 | 512 | ||
BX648041.1 | - | 4 | 574 | |||
DA660812.1 | - | 2 | 318 | |||
DA778416.1 | - | 2 | 317 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
