siRNA Details
Gene Symbol: POTEA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002920.1 | NP_001002920.1 | 8 | 962 | ||
NM_001005365.2 | NP_001005365.2 | 9 | 1100 | |||
XM_011544516.1 | XP_011542818.1 | 8 | 1101 | |||
GenBank mRNA | AY462869.1 | - | 9 | 1100 | ||
AY462870.1 | - | 8 | 962 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
