siRNA Details
Gene Symbol: VWA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022834.4 | NP_073745.2 | 3 | 2201 | ||
NM_199121.2 | NP_954572.2 | 3 | 1806 | |||
GenBank mRNA | AK025868.1 | - | 1 | 1347 | ||
AK096773.1 | - | 3 | 1613 | |||
AL137722.1 | - | 1 | 351 | |||
BC003543.1 | - | 2 | 1485 | |||
BC059409.1 | - | 3 | 2008 | 30349211 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
