siRNA Details
Gene Symbol: CSDE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007553.2 | NP_001007554.1 | 11 | 1622 | ||
NM_001130523.2 | NP_001123995.1 | 11 | 1676 | |||
NM_001242891.1 | NP_001229820.1 | 12 | 1769 | |||
NM_001242892.1 | NP_001229821.1 | 10 | 1235 | |||
NM_001242893.1 | NP_001229822.1 | 9 | 1142 | |||
NM_007158.5 | NP_009089.4 | 10 | 1529 | |||
GenBank mRNA | AB020692.1 | - | 11 | 1536 | ||
AF070542.1 | - | 9 | 1126 | |||
AK022516.1 | - | 9 | 1044 | |||
AK022528.1 | - | 9 | 1044 | |||
AK290146.1 | - | 10 | 1167 | |||
AY049788.1 | - | 8 | 1031 | |||
BC032446.1 | - | 10 | 1464 | 5111213 | ||
BX648561.1 | - | 10 | 1366 | |||
BX648780.1 | - | 10 | 1463 | |||
CR749378.1 | - | 11 | 1626 | |||
CR749431.1 | - | 11 | 1608 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
