siRNA Details
Gene Symbol: CSDE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007553.2 | NP_001007554.1 | 20 | 3239 | ||
NM_001130523.2 | NP_001123995.1 | 20 | 3293 | |||
NM_001242891.1 | NP_001229820.1 | 21 | 3386 | |||
NM_001242892.1 | NP_001229821.1 | 19 | 2852 | |||
NM_001242893.1 | NP_001229822.1 | 18 | 2759 | |||
NM_007158.5 | NP_009089.4 | 19 | 3146 | |||
GenBank mRNA | AB020692.1 | - | 20 | 3153 | ||
AF077054.1 | - | 19 | 3062 | |||
AK022516.1 | - | 18 | 2661 | |||
AK022528.1 | - | 18 | 2661 | |||
AK130392.1 | - | 8 | 1347 | |||
AL050150.1 | - | 9 | 1439 | |||
AL110192.1 | - | 4 | 845 | |||
BC021692.1 | - | 7 | 1149 | |||
BX648561.1 | - | 19 | 2983 | |||
BX648780.1 | - | 19 | 3079 | |||
CR749378.1 | - | 20 | 3243 | |||
CR749431.1 | - | 20 | 3225 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
