siRNA Details
Gene Symbol: CSDE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007553.2 | NP_001007554.1 | 18 | 2682 | ||
NM_001130523.2 | NP_001123995.1 | 18 | 2736 | |||
NM_001242891.1 | NP_001229820.1 | 19 | 2829 | |||
NM_001242892.1 | NP_001229821.1 | 17 | 2295 | |||
NM_001242893.1 | NP_001229822.1 | 16 | 2202 | |||
NM_007158.5 | NP_009089.4 | 17 | 2589 | |||
GenBank mRNA | AB020692.1 | - | 18 | 2596 | ||
AF077054.1 | - | 17 | 2505 | |||
AK022516.1 | - | 16 | 2104 | |||
AK022528.1 | - | 16 | 2104 | |||
AK130392.1 | - | 6 | 790 | |||
AK290146.1 | - | 17 | 2227 | |||
AL050150.1 | - | 7 | 882 | |||
AL110192.1 | - | 2 | 288 | |||
AY049788.1 | - | 15 | 2091 | |||
BC021692.1 | - | 5 | 592 | |||
BC032446.1 | - | 17 | 2524 | 5111213 | ||
BX648561.1 | - | 17 | 2426 | |||
BX648780.1 | - | 17 | 2523 | |||
CR749378.1 | - | 18 | 2686 | |||
CR749431.1 | - | 18 | 2668 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
