siRNA Details
Gene Symbol: OTX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270523.1 | NP_001257452.1 | 5 | 1538 | ||
NM_001270524.1 | NP_001257453.1 | 4 | 1691 | |||
NM_001270525.1 | NP_001257454.1 | 3 | 1485 | |||
NM_021728.3 | NP_068374.1 | 5 | 1562 | |||
NM_172337.2 | NP_758840.1 | 3 | 1461 | |||
NR_073034.1 | 4 | 1386 | ||||
NR_073036.1 | 2 | 1309 | ||||
GenBank mRNA | AB593056.1 | - | 2 | 1312 | ||
AB593057.1 | - | 3 | 1231 | |||
AB593058.1 | - | 5 | 1562 | |||
BC032579.1 | - | 5 | 1563 | 5493541 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
