siRNA Details
Gene Symbol: AQP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317384.1 | NP_001304313.1 | 5 | 2117 | ||
NM_001317387.1 | NP_001304316.1 | 4 | 2036 | |||
NM_001650.5 | NP_001641.1 | 5 | 2117 | |||
NM_004028.4 | NP_004019.1 | 4 | 2244 | |||
XM_011525942.2 | XP_011524244.1 | 5 | 2159 | |||
GenBank mRNA | AB209156.1 | - | 3 | 1650 | ||
AL119338.1 | - | 1 | 21 | |||
R35726.1 | - | 1 | 187 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
