siRNA Details
Gene Symbol: AQP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317384.1 | NP_001304313.1 | 5 | 4438 | ||
NM_001317387.1 | NP_001304316.1 | 4 | 4357 | |||
NM_001650.5 | NP_001641.1 | 5 | 4438 | |||
NM_004028.4 | NP_004019.1 | 4 | 4565 | |||
XM_011525942.2 | XP_011524244.1 | 5 | 4480 | |||
GenBank mRNA | AB209156.1 | - | 3 | 3970 | ||
AK026728.1 | - | 1 | 1709 | |||
BC030745.1 | - | 1 | 436 | |||
BC045780.1 | - | 1 | 816 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
