siRNA Details
Gene Symbol: TMEM179 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286389.1 | NP_001273318.1 | 4 | 2037 | ||
XM_011536745.2 | XP_011535047.1 | 5 | 2764 | |||
XM_011536750.2 | XP_011535052.1 | 5 | 1949 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
