siRNA Details
Gene Symbol: UBC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021009.6 | NP_066289.3 | 2 | 2561 | ||
GenBank mRNA | AB009010.1 | - | 2 | 2171 | ||
AB209782.1 | - | 3975 | ||||
AF052164.1 | - | 2 | 1116 | |||
AK026846.1 | - | 3 | 1042 | |||
AK056939.1 | - | 5 | 1279 | |||
AK092983.1 | - | 2763 | ||||
AK097415.1 | - | 2 | 1843 | |||
AK098827.1 | - | 2 | 2170 | |||
AK129749.1 | - | 2 | 2170 | |||
AK291539.1 | - | 3 | 1942 | |||
AL117514.1 | - | 1 | 765 | |||
BC000449.2 | - | 2 | 2156 | |||
BC008955.2 | - | 3 | 1688 | |||
BC014880.1 | - | 1 | 1082 | |||
BC039193.1 | - | 2 | 2171 | 4076286 | ||
BC080583.1 | - | 2 | 2144 | |||
BC093445.1 | - | 2 | 2170 | |||
BF793240.1 | - | 1 | 678 | |||
M26880.1 | - | 2238 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
