siRNA Details
Gene Symbol: UBC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021009.6 | NP_066289.3 | 2 | 2487 | ||
GenBank mRNA | AB009010.1 | - | 2 | 2097 | ||
AB209782.1 | - | 3901 | ||||
AF052164.1 | - | 2 | 1042 | |||
AK026846.1 | - | 3 | 968 | |||
AK056939.1 | - | 5 | 1205 | |||
AK092983.1 | - | 2689 | ||||
AK097415.1 | - | 2 | 1769 | |||
AK098827.1 | - | 2 | 2096 | |||
AK129749.1 | - | 2 | 2096 | |||
AK291539.1 | - | 3 | 1868 | |||
AL117514.1 | - | 1 | 691 | |||
AY732488.1 | - | 2 | 451 | |||
AY819759.1 | - | 2 | 679 | |||
BC000449.2 | - | 2 | 2082 | |||
BC008955.2 | - | 3 | 1614 | |||
BC014880.1 | - | 1 | 1008 | |||
BC039193.1 | - | 2 | 2097 | 4076286 | ||
BC080583.1 | - | 2 | 2070 | |||
BC093445.1 | - | 2 | 2096 | |||
BF793240.1 | - | 1 | 604 | |||
M17597.1 | - | 1 | 781 | |||
M26880.1 | - | 2164 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
