siRNA Details
Gene Symbol: BRPF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015695.2 | NP_056510.2 | 7 | 2597 | ||
XM_005249010.2 | XP_005249067.1 | 7 | 2479 | |||
XM_005249011.3 | XP_005249068.1 | 7 | 2603 | |||
XM_011514489.1 | XP_011512791.1 | 7 | 2501 | |||
XM_011514490.1 | XP_011512792.1 | 6 | 2399 | |||
XM_011514491.1 | XP_011512793.1 | 6 | 2399 | |||
XM_011514492.1 | XP_011512794.1 | 6 | 2399 | |||
XM_017010742.1 | XP_016866231.1 | 6 | 2399 | |||
XM_017010743.1 | XP_016866232.1 | 4 | 715 | |||
GenBank mRNA | AB033112.1 | - | 7 | 2597 | ||
AK000751.1 | - | 2 | 225 | |||
AK074362.1 | - | 4 | 734 | |||
BC033652.2 | - | 6 | 2316 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
