siRNA Details
Gene Symbol: VSX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256271.1 | NP_001243200.1 | 3 | 816 | ||
NM_001256272.1 | NP_001243201.1 | 3 | 816 | |||
NM_014588.5 | NP_055403.2 | 3 | 816 | |||
NM_199425.2 | NP_955457.1 | 3 | 816 | |||
NR_045948.1 | 3 | 816 | ||||
NR_045951.1 | 3 | 816 | ||||
XM_017027837.1 | XP_016883326.1 | 3 | 533 | |||
XM_017027838.1 | XP_016883327.1 | 3 | 533 | |||
GenBank mRNA | AF176797.2 | - | 3 | 806 | ||
AF251033.1 | - | 3 | 568 | |||
AF251034.1 | - | 3 | 568 | |||
BC126228.1 | - | 3 | 558 | 8991944 | ||
BC136497.1 | - | 3 | 558 | 9020486 | ||
DQ854807.1 | - | 3 | 568 | |||
DQ854808.1 | - | 3 | 568 | |||
DQ854809.1 | - | 3 | 568 | |||
DQ854810.1 | - | 3 | 568 | |||
DQ854811.1 | - | 3 | 568 | |||
DQ854812.1 | - | 3 | 568 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
