siRNA Details
Gene Symbol: HIST1H2BG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003518.3 | NP_003509.1 | 1 | 165 | ||
GenBank mRNA | AK311935.1 | - | 1 | 214 | ||
BC082232.1 | - | 1 | 192 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
